Product Details

SNP ID
rs920485
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125113735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTCCTTCTCCACCAGCTTCTGG[A/G]GTGGCTGGGGGTCTGGAGGCCTGGC
Phenotype
MIM: 613066
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KRT18P59 PubMed Links

Gene Details

Gene
KRT18P59
Gene Name
keratin 18 pseudogene 59
There are no transcripts associated with this gene.

Gene
PKNOX2
Gene Name
PBX/knotted 1 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022062.2 1739 Intron NP_071345.2
XM_005271642.2 1739 Intron XP_005271699.1
XM_005271643.2 1739 Intron XP_005271700.1
XM_006718894.2 1739 Intron XP_006718957.1
XM_011542944.2 1739 Intron XP_011541246.1
XM_011542945.2 1739 Intron XP_011541247.1
XM_011542946.1 1739 Intron XP_011541248.1
XM_011542947.2 1739 Intron XP_011541249.1
XM_017018110.1 1739 UTR 5 XP_016873599.1
XM_017018111.1 1739 Intron XP_016873600.1
XM_017018112.1 1739 Intron XP_016873601.1
Gene
TMEM218
Gene Name
transmembrane protein 218
There are no transcripts associated with this gene.

View Full Product Details