Product Details

SNP ID
rs1063608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95090047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTTATCTTTCTATTCAGCTTAT[C/T]AACTACTGCAGGACTGGCACTACTG
Phenotype
MIM: 606241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DICER1 PubMed Links

Gene Details

Gene
DICER1
Gene Name
dicer 1, ribonuclease III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195573.1 6341 UTR 3 NP_001182502.1
NM_001271282.2 6341 UTR 3 NP_001258211.1
NM_001291628.1 6341 UTR 3 NP_001278557.1
NM_030621.4 6341 UTR 3 NP_085124.2
NM_177438.2 6341 UTR 3 NP_803187.1
XM_011536599.1 6341 UTR 3 XP_011534901.1
XM_011536600.2 6341 UTR 3 XP_011534902.1
XM_011536601.2 6341 UTR 3 XP_011534903.1
XM_011536602.2 6341 UTR 3 XP_011534904.1
XM_011536604.2 6341 UTR 3 XP_011534906.1
XM_011536605.2 6341 UTR 3 XP_011534907.1
XM_017021120.1 6341 UTR 3 XP_016876609.1
XM_017021121.1 6341 UTR 3 XP_016876610.1
XM_017021122.1 6341 UTR 3 XP_016876611.1
XM_017021123.1 6341 UTR 3 XP_016876612.1

View Full Product Details