Product Details

SNP ID
rs1370972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149277347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCATTGCTCAACAGATGATCTG[A/C]GAATCCATTCTGCCTGTGTCTTGCC
Phenotype
MIM: 614410
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AFAP1L1 PubMed Links

Gene Details

Gene
AFAP1L1
Gene Name
actin filament associated protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146337.2 Intron NP_001139809.1
NM_001323062.1 Intron NP_001309991.1
NM_001323063.1 Intron NP_001309992.1
NM_152406.3 Intron NP_689619.1
XM_011537558.1 Intron XP_011535860.1
XM_017009036.1 Intron XP_016864525.1

View Full Product Details