Product Details
- SNP ID
-
rs904546
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:126281650 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTTCCTGAATAAGCTGCAAAAATG[G/A]GCCTTGATCCCTTTGTAGCTCATAA
- Phenotype
-
MIM: 613622
MIM: 606252
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXRED1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8177343,rs8177344] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOXRED1
- Gene Name
- FAD dependent oxidoreductase domain containing 1
There are no transcripts associated with this gene.
- Gene
- TIRAP
- Gene Name
- TIR domain containing adaptor protein
View Full Product Details