Product Details
- SNP ID
-
rs12334564
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:123768584 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TAGGCCATGGGGCAGCCTGGGCCTT[A/C]TGCAGTGTGAGGCGCGGGGCCTCCC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM91A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142309473] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM91A1
- Gene Name
- family with sequence similarity 91 member A1
View Full Product Details