Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022062.2 | 674 | Intron | NP_071345.2 | ||
XM_005271642.2 | 674 | Intron | XP_005271699.1 | ||
XM_005271643.2 | 674 | Intron | XP_005271700.1 | ||
XM_006718894.2 | 674 | Intron | XP_006718957.1 | ||
XM_011542944.2 | 674 | Intron | XP_011541246.1 | ||
XM_011542945.2 | 674 | Intron | XP_011541247.1 | ||
XM_011542946.1 | 674 | Intron | XP_011541248.1 | ||
XM_011542947.2 | 674 | Intron | XP_011541249.1 | ||
XM_017018110.1 | 674 | UTR 5 | XP_016873599.1 | ||
XM_017018111.1 | 674 | Intron | XP_016873600.1 | ||
XM_017018112.1 | 674 | Intron | XP_016873601.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001080546.2 | 674 | Intron | NP_001074015.1 | ||
NM_001258238.1 | 674 | Intron | NP_001245167.1 | ||
NM_001258239.1 | 674 | Intron | NP_001245168.1 | ||
NM_001258240.1 | 674 | Intron | NP_001245169.1 | ||
NM_001258241.1 | 674 | Intron | NP_001245170.1 | ||
NM_001258242.1 | 674 | Intron | NP_001245171.1 | ||
NM_001258243.1 | 674 | Intron | NP_001245172.1 | ||
NM_001258244.1 | 674 | Intron | NP_001245173.1 | ||
NM_001258245.1 | 674 | Intron | NP_001245174.1 | ||
NM_001258246.1 | 674 | Intron | NP_001245175.1 | ||
NM_001258247.1 | 674 | Intron | NP_001245176.1 | ||
XM_006718779.3 | 674 | Intron | XP_006718842.1 | ||
XM_006718780.3 | 674 | Intron | XP_006718843.1 | ||
XM_006718781.3 | 674 | Intron | XP_006718844.1 | ||
XM_006718783.3 | 674 | Intron | XP_006718846.1 | ||
XM_017017322.1 | 674 | Intron | XP_016872811.1 | ||
XM_017017323.1 | 674 | Intron | XP_016872812.1 | ||
XM_017017324.1 | 674 | Intron | XP_016872813.1 |