Product Details

SNP ID
rs3867391
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:158698051 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTATACAATACATTCTTGTTTTGAA[G/T]GGGTGCCTTGTGTCTCCTACTGCCT
Phenotype
MIM: 606639 MIM: 605090
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GFM1 PubMed Links

Gene Details

Gene
GFM1
Gene Name
G elongation factor mitochondrial 1
There are no transcripts associated with this gene.

Gene
RARRES1
Gene Name
retinoic acid receptor responder 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002888.3 Intron NP_002879.2
NM_206963.1 Intron NP_996846.1
XM_005247686.4 Intron XP_005247743.3

View Full Product Details