Product Details

SNP ID
rs802049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87330539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACATTCCCACCACATGAGGGCAAT[C/T]GGGGATTGACAAAGTCCTCTAAAGC
Phenotype
MIM: 616403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TP53TG1 PubMed Links

Gene Details

Gene
TP53TG1
Gene Name
TP53 target 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details