Product Details

SNP ID
rs809203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11761995 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCATTTCGGTCACTGAGGCCCTG[A/T]CCCTTATCCTCCAGGGACATGGTGC
Phenotype
MIM: 600576 MIM: 608933
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C8orf49 PubMed Links

Gene Details

Gene
C8orf49
Gene Name
chromosome 8 open reading frame 49
There are no transcripts associated with this gene.

Gene
GATA4
Gene Name
GATA binding protein 4
There are no transcripts associated with this gene.

Gene
NEIL2
Gene Name
nei like DNA glycosylase 2
There are no transcripts associated with this gene.

View Full Product Details