Product Details
- SNP ID
-
rs1395332
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
21
- Location
-
Chr.1:100038437 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TCCCTCCCTTCCCCCACCCTGTCCG[A/C]GCTGCCGGTGCCTCGGGGTCGCGGA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MFSD14A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115488331] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MFSD14A
- Gene Name
- major facilitator superfamily domain containing 14A
View Full Product Details