Product Details

SNP ID
rs1055004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63390456 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAAACTATTAAGAGCACTGTCT[A/G]TAATTCAAAATCTTTCAAACCCAAT
Phenotype
MIM: 609983
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS4B PubMed Links

Gene Details

Gene
VPS4B
Gene Name
vacuolar protein sorting 4 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004869.3 2114 UTR 3 NP_004860.2
XM_006722582.2 2114 UTR 3 XP_006722645.1
XM_011526268.2 2114 UTR 3 XP_011524570.1

View Full Product Details