Product Details

SNP ID
rs896318
Assay Type
Validated
NCBI dbSNP Submissions
41
Location
Chr.1:205236656 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
TGAGAAGTTGGATAACAGTCAGACA[A/G]AGCACTAGAAGCGATGTCACAAGGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TMCC2 PubMed Links

Gene Details

Gene
TMCC2
Gene Name
transmembrane and coiled-coil domain family 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242925.1 Intron NP_001229854.1
NM_001297611.1 Intron NP_001284540.1
NM_001297613.1 Intron NP_001284542.1
NM_014858.3 Intron NP_055673.2
XM_005245684.1 Intron XP_005245741.1
XM_005245685.4 Intron XP_005245742.1
XM_005245686.3 Intron XP_005245743.1
XM_017003000.1 Intron XP_016858489.1

View Full Product Details