Product Details

SNP ID
rs558924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238430870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAAGGAATCTTGTGAGCAGTA[A/G]GTCTCAACATTGGGCTGAAAATATT
Phenotype
MIM: 605758
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB1 PubMed Links

Gene Details

Gene
ASB1
Gene Name
ankyrin repeat and SOCS box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005246080.1 Intron XP_005246137.1

View Full Product Details