Product Details

SNP ID
rs1131542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172506136 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTACTTGAGAGATGGATTGTTGC[G/T]CTTTCTTCTACAGTCTTTACAAGGA
Phenotype
MIM: 603598
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TNFSF10 PubMed Links

Gene Details

Gene
TNFSF10
Gene Name
tumor necrosis factor superfamily member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190942.1 1325 UTR 3 NP_001177871.1
NM_001190943.1 1325 Intron NP_001177872.1
NM_003810.3 1325 UTR 3 NP_003801.1

View Full Product Details