Product Details

SNP ID
rs1056819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2928631 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCAGGGCTGGAGAGCGCCGGCCA[C/T]GTCCTAGCCTCGGTCCGACTCGTCC
Phenotype
MIM: 602131 MIM: 602631
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHLDA2 PubMed Links

Gene Details

Gene
PHLDA2
Gene Name
pleckstrin homology like domain family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003311.3 562 UTR 3 NP_003302.1
Gene
SLC22A18
Gene Name
solute carrier family 22 member 18
There are no transcripts associated with this gene.

View Full Product Details