Product Details
- SNP ID
-
rs1430597
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:79965699 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAGGGTCACGAGATTTTCTCCTGC[A/G]TCCTCCTCTGCATGCTTTATGGCCT
- Phenotype
-
MIM: 611595
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSBP1L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79685532] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSBP1L1
- Gene Name
- heat shock factor binding protein 1 like 1
- Gene
- TXNL4A
- Gene Name
- thioredoxin like 4A
There are no transcripts associated with this gene.
View Full Product Details