Product Details
- SNP ID
-
hCV8859442
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:6433848 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCAGCAAGGCCAACCCTGAGCCT[C/T]GCTGAAATTGTCAACCGCACCCATG
- Phenotype
-
MIM: 606351
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ESPN
PubMed Links
Gene Details
- Gene
- ESPN
- Gene Name
- espin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031475.2 |
|
Intron |
|
|
NP_113663.2 |
- Gene
- MIR4252
- Gene Name
- microRNA 4252
There are no transcripts associated with this gene.
View Full Product Details