Product Details

SNP ID
rs1050284
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:203128988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGCCACCTTCTGCTTCATCGT[C/G]TCGCTGGCGGTGGCTGATGTGGCCG
Phenotype
MIM: 102775
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000674.2 75 Silent Mutation GTC,GTG V49V NP_000665.1
NM_001048230.1 75 Silent Mutation GTC,GTG V49V NP_001041695.1
XM_005244901.1 75 UTR 5 XP_005244958.1
XM_005244902.4 75 UTR 5 XP_005244959.1
XM_017000342.1 75 UTR 5 XP_016855831.1

View Full Product Details