Product Details

SNP ID
rs1519820
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:78284865 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCTTCTTTGGACAGTCCCAAAAAT[A/G]ACAAGCGATGATTGTGTTTAAGCAA
Phenotype
MIM: 609540
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJA4 PubMed Links

Gene Details

Gene
DNAJA4
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
There are no transcripts associated with this gene.

Gene
WDR61
Gene Name
WD repeat domain 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303247.1 1709 Intron NP_001290176.1
NM_001303248.1 1709 Intron NP_001290177.1
NM_025234.2 1709 Intron NP_079510.1
XM_011522094.1 1709 Intron XP_011520396.1
XM_017022637.1 1709 UTR 3 XP_016878126.1

View Full Product Details