Product Details

SNP ID
rs7572
Assay Type
Validated
NCBI dbSNP Submissions
71
Location
Chr.1:207886664 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
ACCGTCATTGAAACCAGGATCCCTG[T/C]TCAACCCCTCTGGAAAGAAATCAAC
Phenotype
MIM: 142230
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CD34 PubMed Links

Gene Details

Gene
CD34
Gene Name
CD34 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025109.1 2685 UTR 3 NP_001020280.1
NM_001773.2 2685 UTR 3 NP_001764.1

View Full Product Details