Product Details

SNP ID
rs1064539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131429006 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAATCTTCGATGGAACACAGGCC[A/T]CGAGGTTGCTACTGAGAGAAGCTGG
Phenotype
MIM: 610499
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RAPGEF6 PubMed Links

Gene Details

Gene
RAPGEF6
Gene Name
Rap guanine nucleotide exchange factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164386.1 4882 Missense Mutation GAG,GTG E1567V NP_001157858.1
NM_001164387.1 4882 Intron NP_001157859.1
NM_001164388.1 4882 Intron NP_001157860.1
NM_001164389.1 4882 Intron NP_001157861.1
NM_001164390.1 4882 Intron NP_001157862.1
NM_016340.5 4882 Missense Mutation GAG,GTG E1559V NP_057424.3

View Full Product Details