Product Details

SNP ID
rs862703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57353714 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGACTGTGGACTCAGCTGTACT[C/T]ATCATGGCAGAGTTGTGTGACCTTC
Phenotype
MIM: 613840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC2B PubMed Links

Gene Details

Gene
TRAPPC2B
Gene Name
trafficking protein particle complex 2B
There are no transcripts associated with this gene.

Gene
ZNF304
Gene Name
zinc finger protein 304
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290318.1 Intron NP_001277247.1
NM_001290319.1 Intron NP_001277248.1
NM_020657.3 Intron NP_065708.2
XM_011527145.2 Intron XP_011525447.1
XM_017027024.1 Intron XP_016882513.1
Gene
ZNF547
Gene Name
zinc finger protein 547
There are no transcripts associated with this gene.

View Full Product Details