Product Details

SNP ID
rs1014737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41054978 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCAGAGCAATGGATCCTGAAGGG[A/T]TAAGTCCACTGATATAACTTACATT
Phenotype
MIM: 604797
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
APOBEC2 PubMed Links

Gene Details

Gene
APOBEC2
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006789.3 Intron NP_006780.1

View Full Product Details