Product Details

SNP ID
rs28051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:52990007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCCTGCAGCTCCCCAGCGCGGT[C/G]CTTGCTGGCTTTGCAGAAGCCTGCG
Phenotype
MIM: 192974
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITGA2 PubMed Links

Gene Details

Gene
ITGA2
Gene Name
integrin subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002203.3 Intron NP_002194.2

View Full Product Details