Product Details
- SNP ID
-
rs11635335
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:22787334 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTACGCTCGGTAGAGCAAGACCTGG[G/T]ATTTCCAGTCCCCCGCAGGGGCCTC
- Phenotype
-
MIM: 608145
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC283683
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112322285] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC283683
- Gene Name
- uncharacterized LOC283683
There are no transcripts associated with this gene.
- Gene
- NIPA1
- Gene Name
- non imprinted in Prader-Willi/Angelman syndrome 1
View Full Product Details