Product Details

SNP ID
rs1131738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75369538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGGGTTTAGAGCTTGGAGCTCA[A/G]ATGTCAGACAGCAGGAGGAATAAAA
Phenotype
MIM: 154580 MIM: 607776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAN2C1 PubMed Links

Gene Details

Gene
MAN2C1
Gene Name
mannosidase alpha class 2C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256494.1 6445 Intron NP_001243423.1
NM_001256495.1 6445 Intron NP_001243424.1
NM_001256496.1 6445 Intron NP_001243425.1
NM_006715.3 6445 Intron NP_006706.2
XM_005254384.2 6445 Intron XP_005254441.1
XM_017022186.1 6445 Intron XP_016877675.1
XM_017022187.1 6445 Intron XP_016877676.1
XM_017022188.1 6445 Intron XP_016877677.1
Gene
SIN3A
Gene Name
SIN3 transcription regulator family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145357.1 6445 UTR 3 NP_001138829.1
NM_001145358.1 6445 UTR 3 NP_001138830.1
NM_015477.2 6445 UTR 3 NP_056292.1
XM_006720465.3 6445 UTR 3 XP_006720528.1
XM_006720466.3 6445 UTR 3 XP_006720529.1
XM_006720467.3 6445 UTR 3 XP_006720530.1

View Full Product Details