Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286462.1 | Intron | NP_001273391.1 | |||
NM_001286463.1 | Intron | NP_001273392.1 | |||
NM_001286476.1 | Intron | NP_001273405.1 | |||
NM_001286477.1 | Intron | NP_001273406.1 | |||
NM_058180.4 | Intron | NP_478060.2 | |||
XM_005261148.4 | Intron | XP_005261205.1 | |||
XM_006724018.3 | Intron | XP_006724081.1 | |||
XM_011529619.2 | Intron | XP_011527921.1 | |||
XM_011529620.2 | Intron | XP_011527922.1 | |||
XM_011529621.2 | Intron | XP_011527923.1 | |||
XM_011529623.2 | Intron | XP_011527925.1 | |||
XM_011529624.2 | Intron | XP_011527926.1 | |||
XM_011529625.2 | Intron | XP_011527927.1 | |||
XM_011529626.2 | Intron | XP_011527928.1 | |||
XM_011529627.2 | Intron | XP_011527929.1 | |||
XM_011529628.1 | Intron | XP_011527930.1 | |||
XM_017028388.1 | Intron | XP_016883877.1 | |||
XM_017028389.1 | Intron | XP_016883878.1 | |||
XM_017028390.1 | Intron | XP_016883879.1 | |||
XM_017028391.1 | Intron | XP_016883880.1 | |||
XM_017028392.1 | Intron | XP_016883881.1 | |||
XM_017028393.1 | Intron | XP_016883882.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001006114.2 | Intron | NP_001006114.1 | |||
NM_001314022.1 | Intron | NP_001300951.1 | |||
NM_001314023.1 | Intron | NP_001300952.1 | |||
NM_001314024.1 | Intron | NP_001300953.1 | |||
NM_001314025.1 | Intron | NP_001300954.1 | |||
NM_001314026.1 | Intron | NP_001300955.1 | |||
NM_058181.2 | Intron | NP_478061.1 | |||
XM_005261156.3 | Intron | XP_005261213.2 | |||
XM_011529629.2 | Intron | XP_011527931.1 | |||
XM_011529630.2 | Intron | XP_011527932.1 | |||
XM_011529631.2 | Intron | XP_011527933.1 | |||
XM_011529633.2 | Intron | XP_011527935.1 | |||
XM_011529634.2 | Intron | XP_011527936.2 | |||
XM_011529635.2 | Intron | XP_011527937.1 | |||
XM_017028394.1 | Intron | XP_016883883.1 | |||
XM_017028395.1 | Intron | XP_016883884.1 | |||
XM_017028396.1 | Intron | XP_016883885.1 |