Product Details

SNP ID
rs5956229
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:120607086 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCCGTCCAGGTATTTCCTGTGATG[G/T]GTGTAAATGCATTTTTTTTTAAAAT
Phenotype
MIM: 300587
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MCTS1 PubMed Links

Gene Details

Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137554.1 Intron NP_001131026.1
NM_014060.2 Intron NP_054779.1

View Full Product Details