Product Details

SNP ID
rs746424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54023690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTCCCCAAAATATTAGCACCCC[C/T]AAATGATATCCATTTCTGTGTTAGG
Phenotype
MIM: 142974 MIM: 142973 MIM: 142972
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXC4 PubMed Links

Gene Details

Gene
HOXC4
Gene Name
homeobox C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014620.5 Intron NP_055435.2
NM_153633.2 Intron NP_705897.1
Gene
HOXC5
Gene Name
homeobox C5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018953.3 Intron NP_061826.1
Gene
HOXC6
Gene Name
homeobox C6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004503.3 Intron NP_004494.1
NM_153693.4 Intron NP_710160.1

View Full Product Details