Product Details

SNP ID
rs1802581
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:153534763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAAGGTGACATACTCCTGGAAG[C/T]TCACCTCCTGGTCCTTGTTCCGGTC
Phenotype
MIM: 114210 MIM: 176991 MIM: 114110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100A4 PubMed Links

Gene Details

Gene
S100A4
Gene Name
S100 calcium binding protein A4
There are no transcripts associated with this gene.

Gene
S100A5
Gene Name
S100 calcium binding protein A5
There are no transcripts associated with this gene.

Gene
S100A6
Gene Name
S100 calcium binding protein A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014624.3 520 Missense Mutation NP_055439.1
XM_017002033.1 520 Missense Mutation XP_016857522.1

View Full Product Details