Product Details

SNP ID
rs889391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70017728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTGAGTTTTCATTTTGGGAAGA[A/G]GAGTGTGCCCAAGGCCTTGGGCAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDXDC2P PubMed Links

Gene Details

Gene
PDXDC2P
Gene Name
pyridoxal dependent decarboxylase domain containing 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details