Product Details
- SNP ID
-
rs7659
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:195568790 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ACAGGGTAGGGCATGGTTACATGTT[C/T]AGGTCAACTTCCTTTGTCGTGGTTG
- Phenotype
-
MIM: 107740
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APOD
PubMed Links
Gene Details
- Gene
- APOD
- Gene Name
- apolipoprotein D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001647.3 |
1042 |
UTR 3 |
|
|
NP_001638.1 |
View Full Product Details