Product Details

SNP ID
rs1050153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196049444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTAAAGGTCATGCAAGTTCTA[A/G]AATAGTGAATCATGACAGAACTCAT
Phenotype
MIM: 190010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFRC PubMed Links

Gene Details

Gene
TFRC
Gene Name
transferrin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128148.2 4525 UTR 3 NP_001121620.1
NM_001313965.1 4525 UTR 3 NP_001300894.1
NM_001313966.1 4525 UTR 3 NP_001300895.1
NM_003234.3 4525 UTR 3 NP_003225.2

View Full Product Details