Product Details
- SNP ID
-
rs2295566
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23566304 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCCAGGGCAGACACCCATGAATGTA[A/T]CCCTATTTCCAGATGCACTTCCCAC
- Phenotype
-
MIM: 616536
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CST9L
PubMed Links
Gene Details
- Gene
- CST9L
- Gene Name
- cystatin 9-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080610.2 |
|
Intron |
|
|
NP_542177.1 |
View Full Product Details