Product Details

SNP ID
rs35551926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:35234492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATAAATTAGATTCAGAATACA[A/G]CAAATCAAAATTTACAATAAGAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANP32AP1 PubMed Links

Gene Details

Gene
ANP32AP1
Gene Name
acidic nuclear phosphoprotein 32 family member A pseudogene 1
There are no transcripts associated with this gene.

Gene
DPH6
Gene Name
diphthamine biosynthesis 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141972.1 Intron NP_001135444.1
NM_080650.3 Intron NP_542381.1
XM_011522160.1 Intron XP_011520462.1
XM_017022707.1 Intron XP_016878196.1
XM_017022708.1 Intron XP_016878197.1
XM_017022709.1 Intron XP_016878198.1

View Full Product Details