Product Details

SNP ID
rs2015
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:38878729 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCCCCTACGCTGGGTGTGGGTTA[G/T]ATTCAGAGGCCCACACCCACACTGG
Phenotype
MIM: 604480
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RINL PubMed Links

Gene Details

Gene
RINL
Gene Name
Ras and Rab interactor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195833.1 1783 Intron NP_001182762.1
NM_198445.3 1783 Intron NP_940847.1
XM_006723015.2 1783 Intron XP_006723078.1
XM_006723016.2 1783 Intron XP_006723079.1
XM_011526454.2 1783 Intron XP_011524756.1
XM_011526455.2 1783 Intron XP_011524757.1
Gene
SIRT2
Gene Name
sirtuin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193286.1 1783 UTR 3 NP_001180215.1
NM_012237.3 1783 UTR 3 NP_036369.2
NM_030593.2 1783 UTR 3 NP_085096.1
XM_006723111.1 1783 UTR 3 XP_006723174.1
XM_011526654.1 1783 UTR 3 XP_011524956.1
XM_011526655.1 1783 UTR 3 XP_011524957.1
XM_017026500.1 1783 UTR 3 XP_016881989.1

View Full Product Details