Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001292018.1 | 3653 | UTR 3 | NP_001278947.1 | ||
NM_001292019.1 | 3653 | UTR 3 | NP_001278948.1 | ||
NM_004762.3 | 3653 | UTR 3 | NP_004753.1 | ||
NM_017456.3 | 3653 | UTR 3 | NP_059430.2 | ||
XM_011525475.2 | 3653 | UTR 3 | XP_011523777.1 | ||
XM_011525476.2 | 3653 | UTR 3 | XP_011523778.1 | ||
XM_011525477.1 | 3653 | UTR 3 | XP_011523779.1 | ||
XM_011525478.1 | 3653 | UTR 3 | XP_011523780.1 | ||
XM_017025336.1 | 3653 | UTR 3 | XP_016880825.1 | ||
XM_017025337.1 | 3653 | Intron | XP_016880826.1 | ||
XM_017025338.1 | 3653 | UTR 3 | XP_016880827.1 |