Product Details

SNP ID
rs4520105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130171436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAATTTAATGTGTTTTTAACTCA[A/G]GAATCTGAGAGAATGTATGATTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM209 PubMed Links

Gene Details

Gene
TMEM209
Gene Name
transmembrane protein 209
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301163.1 Intron NP_001288092.1
NM_032842.3 Intron NP_116231.2
XM_005250655.1 Intron XP_005250712.1
XM_011516646.2 Intron XP_011514948.1

View Full Product Details