Product Details
- SNP ID
-
rs3010876
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
71
- Location
-
Chr.1:12719558 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGATCTGCCTCCGCTAAAGTATGA[C/T]CCCGATGTTGTGGTCACGGATTTCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AADACL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78912772] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AADACL3
- Gene Name
- arylacetamide deacetylase like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001103170.2 |
314 |
Silent Mutation |
GAC,GAT |
D84D |
NP_001096640.2 |
View Full Product Details