Product Details

SNP ID
rs3010877
Assay Type
Functionally tested
NCBI dbSNP Submissions
90
Location
Chr.1:12719616 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAATCCCTGTGAAGCTGTACCAA[C/T]CCAAGGCATCCACCTGCACCCTGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AADACL3 PubMed Links

Gene Details

Gene
AADACL3
Gene Name
arylacetamide deacetylase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103170.2 372 Missense Mutation CCC,TCC P104S NP_001096640.2

View Full Product Details