Product Details

SNP ID
rs7560684
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:106066249 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCCATTCCCTTCCAGGCAGTCGCTT[A/G]GCAACCAAGTTGACTTGCGCTTCCC
Phenotype
MIM: 611752
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf40 PubMed Links

Gene Details

Gene
C2orf40
Gene Name
chromosome 2 open reading frame 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032411.2 Intron NP_115787.1
XM_006712799.3 Intron XP_006712862.1
XM_011511993.2 Intron XP_011510295.1
XM_017005106.1 Intron XP_016860595.1

View Full Product Details