Product Details

SNP ID
rs9835936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186148454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGAGAGAACCTCTGGGAAGCAG[A/C]ATGAGGCGCTCGTTTTCTTGTGTGG
Phenotype
MIM: 601854
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DGKG PubMed Links

Gene Details

Gene
DGKG
Gene Name
diacylglycerol kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080744.1 4432 UTR 3 NP_001074213.1
NM_001080745.1 4432 UTR 3 NP_001074214.1
NM_001346.2 4432 UTR 3 NP_001337.2

View Full Product Details