Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135565.1 | Intron | NP_001129037.1 | |||
NM_001178135.1 | Intron | NP_001171606.1 | |||
NM_001178136.1 | Intron | NP_001171607.1 | |||
NM_012080.4 | Intron | NP_036212.3 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000351.5 | Intron | NP_000342.2 | |||
NM_001320750.1 | Intron | NP_001307679.1 | |||
NM_001320751.1 | Intron | NP_001307680.1 | |||
NM_001320752.1 | Intron | NP_001307681.1 | |||
NM_001320753.1 | Intron | NP_001307682.1 | |||
NM_001320754.1 | Intron | NP_001307683.1 |