Product Details

SNP ID
rs11923882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119432511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTTAGATTATATTTTAGGTA[C/T]ATATTTTATTTAGGATAAACACAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM39A PubMed Links

Gene Details

Gene
TMEM39A
Gene Name
transmembrane protein 39A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018266.2 Intron NP_060736.1
XM_005247578.1 Intron XP_005247635.1
XM_006713687.1 Intron XP_006713750.1
XM_011512958.2 Intron XP_011511260.1
XM_017006788.1 Intron XP_016862277.1

View Full Product Details