Product Details

SNP ID
rs1196352
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:151629256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTCATCCCCCTATTGACTTTTT[A/G]TAAAAACAAATTTGGAAACAACCTA
Phenotype
MIM: 611541
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNX27 PubMed Links

Gene Details

Gene
SNX27
Gene Name
sorting nexin family member 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030918.5 Intron NP_112180.4
XM_005245509.2 Intron XP_005245566.1
XM_005245510.3 Intron XP_005245567.1
XM_005245511.4 Intron XP_005245568.1
XM_011510024.2 Intron XP_011508326.1
XM_011510025.2 Intron XP_011508327.1
XM_011510026.2 Intron XP_011508328.1
XM_017002417.1 Intron XP_016857906.1

View Full Product Details