Product Details

SNP ID
rs3814880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30006574 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCCCCTCCCTGGCCTCCCTCCA[T/C]GTCCCGTCCCCTCCTGGGTACCTCG
Phenotype
MIM: 604567
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DOC2A PubMed Links

Gene Details

Gene
DOC2A
Gene Name
double C2 domain alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282062.1 Intron NP_001268991.1
NM_001282063.1 Intron NP_001268992.1
NM_001282068.1 Intron NP_001268997.1
NM_003586.2 Intron NP_003577.2
XM_011545975.1 Intron XP_011544277.1
XM_017023776.1 Intron XP_016879265.1
XM_017023777.1 Intron XP_016879266.1
XM_017023778.1 Intron XP_016879267.1
Gene
INO80E
Gene Name
INO80 complex subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304562.1 Intron NP_001291491.1
NM_001304563.1 Intron NP_001291492.1
NM_173618.2 Intron NP_775889.1
XM_011545809.2 Intron XP_011544111.1
XM_011545811.2 Intron XP_011544113.1
XM_011545812.2 Intron XP_011544114.1
XM_017023169.1 Intron XP_016878658.1

View Full Product Details