Product Details

SNP ID
rs241419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32856171 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGGCAGTGGAGTTTGACGGGGGC[A/G]TTGTGATGGGTTCTGATTCCCGAGT
Phenotype
MIM: 177045 MIM: 170260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMB8-AS1 PubMed Links

Gene Details

Gene
PSMB8-AS1
Gene Name
PSMB8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
PSMB9
Gene Name
proteasome subunit beta 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002800.4 163 Missense Mutation ATT,GTT I32V NP_002791.1
Gene
TAP1
Gene Name
transporter 1, ATP binding cassette subfamily B member
There are no transcripts associated with this gene.

View Full Product Details