Product Details

SNP ID
rs430083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34348177 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCTCACTGTTGGTGACAGGGGA[C/T]GGGGGCCCTTGCCTCAGTGGGGCTG
Phenotype
MIM: 606471 MIM: 608963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOP10 PubMed Links

Gene Details

Gene
NOP10
Gene Name
NOP10 ribonucleoprotein
There are no transcripts associated with this gene.

Gene
NUTM1
Gene Name
NUT midline carcinoma family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284292.1 691 Silent Mutation GAC,GAT D103D NP_001271221.1
NM_001284293.1 691 Silent Mutation GAC,GAT D93D NP_001271222.1
NM_175741.2 691 Silent Mutation GAC,GAT D75D NP_786883.1

View Full Product Details