Product Details

SNP ID
rs496798
Assay Type
Functionally Tested
NCBI dbSNP Submissions
40
Location
Chr.1:100042434 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAGGAGTCAGCAACCGTTTCAA[A/G]AGCAGGACCCAAGCTCTGACCCTTT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MFSD14A PubMed Links

Gene Details

Gene
MFSD14A
Gene Name
major facilitator superfamily domain containing 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033055.2 Intron NP_149044.2
XM_005271118.1 Intron XP_005271175.1
XM_011541957.1 Intron XP_011540259.1
XM_017002084.1 Intron XP_016857573.1

View Full Product Details