Product Details

SNP ID
rs312463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7013230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTAGGAGAAACAAGAGCGGGGT[G/T]CCTGAGAGGTGCTTGAGTTGAATGG
Phenotype
MIM: 152391 MIM: 608992 MIM: 610718 MIM: 615672
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALOX12 PubMed Links

Gene Details

Gene
ALOX12
Gene Name
arachidonate 12-lipoxygenase, 12S type
There are no transcripts associated with this gene.

Gene
ALOX12-AS1
Gene Name
ALOX12 antisense RNA 1
There are no transcripts associated with this gene.

Gene
BCL6B
Gene Name
B-cell CLL/lymphoma 6B
There are no transcripts associated with this gene.

Gene
C17orf49
Gene Name
chromosome 17 open reading frame 49
There are no transcripts associated with this gene.

Gene
MIR195
Gene Name
microRNA 195
There are no transcripts associated with this gene.

Gene
MIR497
Gene Name
microRNA 497
There are no transcripts associated with this gene.

Gene
MIR497HG
Gene Name
mir-497-195 cluster host gene
There are no transcripts associated with this gene.

Gene
RNASEK
Gene Name
ribonuclease K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004333.4 Intron NP_001004333.2
Gene
RNASEK-C17orf49
Gene Name
RNASEK-C17orf49 readthrough
There are no transcripts associated with this gene.

View Full Product Details